missing translation for 'onlineSavingsMsg'
Learn More

Lamivudine 98.0+%, TCI America™

Catalog No. L0217100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0217100MG 100 mg
L02171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0217100MG Supplier TCI America Supplier No. L0217100MG

Chemical Identifiers

CAS 134678-17-4
Molecular Formula C8H11N3O3S
Molecular Weight (g/mol) 229.254
MDL Number MFCD00869739
InChI Key JTEGQNOMFQHVDC-NKWVEPMBSA-N
Synonym lamivudine, epivir, zeffix, heptovir, epivir-hbv, hepitec, heptodin, 2',3'-dideoxy-3'-thiacytidine, lamivir, 3tc
PubChem CID 60825
ChEBI CHEBI:63577
IUPAC Name 4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one
SMILES C1C(OC(S1)CO)N2C=CC(=NC2=O)N

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 100 mg
Formula Weight 229.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Lamivudine
RTECSNumber UW7361333
TSCA No
Recommended Storage Freezer