missing translation for 'onlineSavingsMsg'
Learn More

Lamivudine, 98%

Catalog No. AC461090010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461090010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC461090010 Supplier Thermo Scientific Chemicals Supplier No. 461090010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 134678-17-4 | C8H11N3O3S | 229.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 134678-17-4
Molecular Formula C8H11N3O3S
Molecular Weight (g/mol) 229.26
InChI Key JTEGQNOMFQHVDC-NKWVEPMBSA-N
Synonym lamivudine, epivir, zeffix, heptovir, epivir-hbv, hepitec, heptodin, 2',3'-dideoxy-3'-thiacytidine, lamivir, 3tc
PubChem CID 60825
ChEBI CHEBI:63577
IUPAC Name 4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one
SMILES C1C(OC(S1)CO)N2C=CC(=NC2=O)N

Specifications

Melting Point 160.0°C to 162.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: 70g/L (20°C).
Formula Weight 229.26
Percent Purity 98%
Chemical Name or Material Lamivudine
Hazard Category Reproductive toxicity (category 2), Specific target organ toxicity after repeated exposure (category 2)
Hazard Statement Suspected of damaging fertility or the unborn child, May cause damage to organs through prolonged or repeated exposure

RUO – Research Use Only