missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Laminating Pouches, Header: CAUTION
Description
- Made of durable polyester
- Available clear or with OSHA headers
- Have holes in each corner for easy mounting
- Pouch size: 10 mil. thick (5 mil./side)
Specifications
Specifications
| Product Type | Laminating Pouch |
| Color | Clear |
| For Use With (Equipment) | Brady Label Printers |
| Material | Polyester (B-855) |
| Dimensions (L x W) | 20.3 x 27.9 cm (W x H) |
| Description | Holes Included |
| Shape | Rectangle |