missing translation for 'onlineSavingsMsg'
Learn More

Laminarin

Catalog No. AAJ66193MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66193MD 250 mg
AAJ6619303 1 g
AAJ6619306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ66193MD Supplier Thermo Scientific Chemicals Supplier No. J66193MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9008-22-4 | C18H32O16 | 504.438 g/mol

Laminarin is a storage glucan and is now used to study it’s interaction with laminarinase as its substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Laminarin is a storage glucan and is now used to study it’s interaction with laminarinase as its substrate.

Solubility
Soluble in dimethyl sulfoxide (slightly, heated). Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 9008-22-4
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
MDL Number MFCD00046691
InChI Key DBTMGCOVALSLOR-DEVYUCJPSA-N
Synonym laminarin, laminaran
PubChem CID 46173707
IUPAC Name (2S,3R,4S,5R,6R)-4-[(2S,3R,4S,5R,6R)-3,5-dihydroxy-6-(hydroxymethyl)-4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,5-triol
SMILES C(C1C(C(C(C(O1)O)O)OC2C(C(C(C(O2)CO)O)OC3C(C(C(C(O3)CO)O)O)O)O)O)O

Specifications

Color White
Quantity 250 mg
Merck Index 14,5350
Solubility Information Soluble in dimethyl sulfoxide (slightly,heated). Slightly soluble in water.
Optical Rotation −45° (c=2 in Water)
Physical Form Powder
Chemical Name or Material Laminarin
EINECSNumber 232-712-4
RTECSNumber OE1523500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only