missing translation for 'onlineSavingsMsg'
Learn More

Laminaran from Eisenia Bicyclis, TCI America™

Catalog No. L00881G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L00881G 1 g
L008825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L00881G Supplier TCI America Supplier No. L00881G

Chemical Identifiers

CAS 9008-22-4
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
MDL Number MFCD00466918
InChI Key DBTMGCOVALSLOR-DEVYUCJPSA-N
Synonym laminarin, laminaran
PubChem CID 46173707
IUPAC Name (2S,3R,4S,5R,6R)-4-[(2S,3R,4S,5R,6R)-3,5-dihydroxy-6-(hydroxymethyl)-4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,5-triol
SMILES C(C1C(C(C(C(O1)O)O)OC2C(C(C(C(O2)CO)O)OC3C(C(C(C(O3)CO)O)O)O)O)O)O

Specifications

Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Laminaran from Eisenia Bicyclis
EINECSNumber (8)-0073
TSCA Yes