missing translation for 'onlineSavingsMsg'
Learn More

lambda-Carrageenan (Low-viscosity), TCI America™

Catalog No. c28711g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C28711G Supplier TCI America Supplier No. C28711G
Only null left

Chemical Identifiers

CAS 9064-57-7
Molecular Formula C12H19O20S3-3
Molecular Weight (g/mol) 579.444
MDL Number MFCD00151513
InChI Key UWPXLSAITSWCRB-UHFFFAOYSA-K
PubChem CID 91972149
IUPAC Name [5-[4,5-dihydroxy-6-(hydroxymethyl)-3-sulfonatooxyoxan-2-yl]oxy-2,4-dihydroxy-6-(sulfonatooxymethyl)oxan-3-yl] sulfate
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)OS(=O)(=O)[O-])O)COS(=O)(=O)[O-])OS(=O)(=O)[O-])O)O)O

Specifications

Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material lambda-Carrageenan (Low-viscosity)
RTECSNumber FI0704000
TSCA No