Learn More
ACROBiosystems Inc Recombinant Protein;Cynomolgus LAG-3 / CD223 Protein, Fc Tag;HEK293;100UG;LA3,LAG3,CD223,LAG-3,FDC
Catalog No. 501657878
Encompass
Recombinant Protein;Cynomolgus LAG-3 / CD223 Protein, Fc Tag;100UG;Cynomolgus LAG-3, Fc Tag (LA3-C5252) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - His 449 (Accession # XP_005570011.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.0 kDa. The protein migrates as 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus LAG-3, Fc Tag (Cat. No. LA3-C5252) at 2 g/mL (100 L/well) can bind Anti-Human LAG3 MAb, Human IgG1 with a linear range of 8-31 ng/mL (QC tested).;Flow Cytometry assay shows thatCynomolgus LAG-3, FcTag (Cat. No. LA3-C5252) can bind to daudi cell surface. The concentration of LAG-3 used is 1 g/ml (Routinely tested).
Catalog No. 501657878 Supplier ACROBiosystems Inc Supplier No. LA3C5252100UG
Recombinant Protein;Cynomolgus LAG-3 / CD223 Protein, Fc Tag;100UG;Cynomolgus LAG-3, Fc Tag (LA3-C5252) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - His 449 (Accession # XP_005570011.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.0 kDa. The protein migrates as 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus LAG-3, Fc Tag (Cat. No. LA3-C5252) at 2 g/mL (100 L/well) can bind Anti-Human LAG3 MAb, Human IgG1 with a linear range of 8-31 ng/mL (QC tested).;Flow Cytometry assay shows thatCynomolgus LAG-3, FcTag (Cat. No. LA3-C5252) can bind to daudi cell surface. The concentration of LAG-3 used is 1 g/ml (Routinely tested).