missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Laemmli SDS sample buffer, non-reducing (4X)

Catalog No. AAJ63615AD
Change view
Click to view available options
Quantity:
25 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63615AD 50 mL
AAJ63615AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63615AD Supplier Thermo Scientific Chemicals Supplier No. J63615AD

Laemmli SDS sample buffer is used to prepare protein samples in the Laemmli SDS-PAGE system.

Laemmli SDS sample buffer is used for protein sample preparation to be used in the Laemmli SDS-PAGE system, The SDS detergent denatures the proteins and subunits and gives each an overall negative charge so that each will separate based on size. The bromophenol blue serves as a dye front that runs ahead of the proteins and also serves to make it easier to see the sample during loading.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Laemmli SDS sample buffer is composed of several compounds, including 375 mM Tris-HCl (pH 6.8), 9% SDS, 50% glycerol, 9% β-mercaptoethanol, 0.03% bromophenol blue
  • The SDS compound present in the Laemmli SDS sample buffer binds noncovalently to proteins. SDS is negatively charged and can mask the intrinsic charge of the protein it binds

Application

  • Laemmli SDS sample buffer is used to denature proteins and for protein sample preparation to be used in the SDS-PAGE system
  • This sample buffer is used as an electrophoretic dye for denaturation of proteins and monitoring the front of the running gel
  • It is also used for polyacrylamide protein gel analysis

Specifications

Color Blue
Concentration 4X
Form Liquid
For Use With (Application) Laemmli SDS-PAGE
Quantity 50 mL
Product Type Sample Loading Buffer

RUO – Research Use Only