missing translation for 'onlineSavingsMsg'
Learn More
Learn More
National Safety Apparel™ Enespro™ Ladies Coverall 7 oz. Navy Ultrasoft Zipper Closure 32 in. Inseam
Durable and comfortable ultrasoft coveralls offer a unique blend of 88% cotton and 12% high-tenacity nylon for a smooth feel. The inherently FR material delivers exceptional performance and reliability.
Supplier: National Safety Apparel™ C88UJWMD32
Description
- Comfortable sewn-in elastic waist for better fit
- Dual certified to protect against flash fire and arc flash hazards
- 2-way covered zipper front for easy entry
- Two front chest pockets with flaps and hidden snap closures
- Large reinforced back pockets
- Convenient 6 pocket style with side pass through access
- Leg openings made to fit over work boots
- All metal covered to ensure no contact with skin
- Action back allows for full range of motion
- FR fabric will not melt, drip, or ignite
- Home laundry
- Made in the USA
Specifications
| Coveralls | |
| Women's | |
| Navy | |
| Elastic (Stitched-in) | |
| Collared | |
| Covered Zipper | |
| Non-sterile | |
| Flame | |
| NFPA 70E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269 | |
| Coveralls |
| Medium | |
| Cotton, Nylon | |
| Reusable | |
| Open | |
| Unhooded | |
| Stitched | |
| CAT 2 | |
| 8 cal/cm² | |
| None |