missing translation for 'onlineSavingsMsg'
Learn More

Lactulose, Powder, 99%, Spectrum™ Chemical

Catalog No. p-7165803
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
5 kg
500 g
Packaging:
Amber Glass Bottle
Poly Pail
Catalog No. Quantity Packaging
18611189 5 g Amber Glass Bottle
18611188 25 g Amber Glass Bottle
18611187 100 g Amber Glass Bottle
18611191 500 g Amber Glass Bottle
18611190 5 kg Poly Pail
5 options

Catalog No. 18611189

Supplier: Spectrum Chemical Mfg Cor L12475GM

Only null left

Chemical Identifiers

CAS 4618-18-2

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.