missing translation for 'onlineSavingsMsg'
Learn More

Lactulose, Powder, 99%, Spectrum™ Chemical

Catalog No. 18611189
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611189 5 g Amber Glass Bottle
18611188 25 g Amber Glass Bottle
18611187 100 g Amber Glass Bottle
18611191 500 g Amber Glass Bottle
18611190 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611189 Supplier Spectrum Chemical Mfg Cor Supplier No. L12475GM
Only null left

Chemical Identifiers

CAS 4618-18-2

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.