missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lactose octaacetate, 98%

Catalog No. AAJ6669506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6669506 5 g
AAJ6669514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6669506 Supplier Thermo Scientific Chemicals Supplier No. J6669506

CAS: 6291-42-5 | C28H38O19 | 678.593 g/mol

Lactose octaacetate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lactose octaacetate is used as pharmaceutical intermediates.

Solubility
Soluble in chloroform

Notes
Protect from heat. Incompatible with heat and oxidizing agents.

Chemical Identifiers

CAS 6291-42-5
Molecular Formula C28H38O19
Molecular Weight (g/mol) 678.593
MDL Number MFCD00064040
InChI Key WOTQVEKSRLZRSX-SJSKHYGQSA-N
Synonym lactose octaacetate, 2r,3r,4s,5r,6s-4,5,6-tris acetyloxy-3-2r,3r,4s,5s,6r-3,4,5-tris acetyloxy-6-acetyloxy methyl oxan-2-yl oxy oxan-2-yl methyl acetate, 4-o-2-o,3-o,4-o,6-o-tetraacetyl-alpha-d-galactopyranosyl-beta-d-glucopyranose tetraacetate
PubChem CID 11767585
IUPAC Name [(2R,3R,4S,5R,6S)-4,5,6-triacetyloxy-3-[(2R,3R,4S,5S,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC2C(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in chloroform
Formula Weight 678.59
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Lactose octaacetate
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only