missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactose, Monohydrate, Spray-Dried Powder, NF, Spectrum™ Chemical
LA105, 64044-51-5, C12H22O11·H2O
$353.94 - $5,491.38
Chemical Identifiers
| CAS | 64044-51-5 |
|---|---|
| Molecular Formula | C12H24O12 |
| Molecular Weight (g/mol) | 360.31 |
| InChI Key | WSVLPVUVIUVCRA-RJMJUYIDSA-N |
| IUPAC Name | (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate |
| SMILES | O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18603698
|
N/A | 500 g | Poly Bottle |
Each for $353.94
|
|
||||
|
18603699
|
N/A | 2.5 kg | Poly Pail |
Each for $1,285.30
|
|
||||
|
18603700
|
N/A | 12 kg | Poly Pail |
Each for $2,361.94
|
|
||||
|
18603701
|
N/A | 25 kg | Fiber Drum |
Each for $5,491.38
|
|
||||
Description
Spectrum™ Chemical Lactose, Monohydrate, Spray-Dried Powder, NF is used to help form tablets because it has excellent compressibility properties and as a diluent powder for dry-powder inhalations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 64044-51-5 | |
| 360.31 | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate |
| C12H24O12 | |
| WSVLPVUVIUVCRA-RJMJUYIDSA-N | |
| O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O |
Specifications
| 64044-51-5 | |
| 0.1% | |
| C12H24O12 | |
| WSVLPVUVIUVCRA-RJMJUYIDSA-N | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate | |
| NF |
| 100% | |
| 1% | |
| 110.3° | |
| O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O | |
| 360.31 |