missing translation for 'onlineSavingsMsg'
Learn More

Lactose, Monohydrate, Powder, NF, Spectrum™ Chemical

LA106, 64044-51-5, C12H22O11·H2O

Supplier:  Spectrum Chemical Mfg Cor LA1062.5KG

Quantity 2.5 kg
Packaging Poly Pail

 View more versions of this product

Catalog No. 18603695


Only null left

Description

Description

Spectrum™ Chemical Lactose, Monohydrate, Powder, NF is used to help form tablets because it has excellent compressibility properties and as a diluent powder for dry-powder inhalations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

64044-51-5
360.31
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate
C12H24O12
WSVLPVUVIUVCRA-RJMJUYIDSA-N
O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

Specifications

64044-51-5
0.1%
Poly Pail
2.5 kg
WSVLPVUVIUVCRA-RJMJUYIDSA-N
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate
NF
100%
0.5%
C12H24O12
+54.4° to +55.9°
O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O
360.31
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions