missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactose, Monohydrate, BiotechGrade, Spectrum™ Chemical
Description
Spectrum™ Chemical Lactose, Monohydrate, BiotechGrade , also known as milk sugar, is a disaccharide sugar and is added to pills as a filler because of its physical properties. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.
Specifications
Specifications
| Quantity | 12 kg |
| Grade | Biotech |
| Loss on Drying | 0.5% |
| Packaging | Poly Pail |
| Appearance | Fine, white powder |