Learn More
MilliporeSigma™ Lactophenol Blue Solution
Description
Applications
- Fungi appear dark blue
- Stain air-dried, heat-fixed smears of bacteriological material, e.g. sputum, FNAB, lavages, imprints, body fluids, puss, exudates, liquid solid cultures
Specifications
Specifications
| Quantity | 100 mL |
| Packaging | Glass Bottle |