missing translation for 'onlineSavingsMsg'
Learn More

Lactobionic Acid, EP, 98-102%, Spectrum™ Chemical

Catalog No. 18603689
Change view
Click to view available options
Quantity:
5 kg
Packaging:
Poly Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603689 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18603689 Supplier Spectrum Chemical Mfg Cor Supplier No. L16045KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L1604, 96-82-2, C12H22O12

Spectrum™ Chemical Lactobionic Acid, EP is also known as Galactosylgluconic acid. It is a food additive used as a stabilizer and in the pharmaceutical industry as an excipient for formulation. The EP grade indicates it is grade suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 96-82-2
Molecular Formula C12H22O12
Molecular Weight (g/mol) 358.30
InChI Key JYTUSYBCFIZPBE-AMTLMPIISA-N
IUPAC Name (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-{[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanoic acid
SMILES OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C(O)=O

Specifications

CAS Max % 100%
Packaging Poly Pail
Quantity 5 kg
Specific Rotation +23.0° to +29.0°
Percent Purity 98 to 102%
Grade Multi Compendia