missing translation for 'onlineSavingsMsg'
Learn More

Lactobionic acid, 97%

Catalog No. AAJ6636830
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6636830 250 g
AAJ6636814 25 g
AAJ6636822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6636830 Supplier Thermo Scientific Chemicals Supplier No. J6636830

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96-82-2 | C12H22O12 | 358.296 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96-82-2
Molecular Formula C12H22O12
Molecular Weight (g/mol) 358.296
MDL Number MFCD00078147
InChI Key JYTUSYBCFIZPBE-AMTLMPIISA-N
Synonym lactobionic acid, 4-o-beta-d-galactopyranosyl-d-gluconic acid, unii-65r938s4dv, 4-beta-d-galactosido-d-gluconic acid, dsstox_cid_28787, dsstox_rid_83056, dsstox_gsid_48861, lactobionicacid, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-hexanoic acid, prestwick_560
PubChem CID 7314
ChEBI CHEBI:55481
IUPAC Name (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanoic acid
SMILES C(C1C(C(C(C(O1)OC(C(CO)O)C(C(C(=O)O)O)O)O)O)O)O

Specifications

Melting Point 113°C to 118°C
Color White
Odor Odorless
Quantity 250 g
Merck Index 14,5342
Solubility Information Soluble in water
Optical Rotation +25° (c=10 in Water)
Formula Weight 358.3
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Lactobionic acid
Show More Show Less
EINECSNumber 202-538-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only