missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid (USP/FCC), Fisher Chemical

Catalog No. A159500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A159500 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A159500 Supplier Thermo Fisher Scientific Supplier No. BPA159500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Chemical Identifiers

CAS 50-21-5 , 97-73-4
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

pH 2,8
Melting Point 18°C
Boiling Point 122°C
Density 1.200g/cm3
Quantity 500 mL
Ignition Residue 0.05% max. (USP), 0.05% max. (USP), 0.1% max. (FCC), 0.1% max. (FCC)
Packaging Glass Bottle
Physical Form Viscous Liquid
Linear Formula CH3CH(OH)CO2H
UN Number 3265
Formula Weight 90.08
Percent Purity 95.0 to 105.0% (FCC)
Grade FCC/USP
Chemical Name or Material Lactic Acid
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation Class 8: Corrosive