missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactic Acid, Racemic, USP, 88-92%, Spectrum™ Chemical
Description
Spectrum™ Chemical Lactic Acid, Racemic, USP is used as a food preservative, curing agent, and flavoring agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| UN Number | UN3265 |
| Percent Purity | 88 to 92% |
| Grade | USP |
| Quantity | 4 L |
| Ignition Residue | 0.05% |
| Packaging | Poly Bottle |