missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, Racemic, USP, 88-92%, Spectrum™ Chemical

Catalog No. 18603679
Change view
Click to view available options
Quantity:
125 mL
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603679 125 mL Amber Glass Bottle
18603680 500 mL Poly Bottle
18603681 6 x 500 mL Poly Bottle
18603682 4 L Amber Glass Bottle
18603685 4 L Poly Bottle
18603683 4 x 4 L Amber Glass Bottle
18603686 4 x 4 L Poly Bottle
18603684 20 L Poly Drum
18603687 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18603679 Supplier Spectrum Chemical Mfg Cor Supplier No. L1010125ML
Only null left

L1010, 50-21-5, C3H6O3

Spectrum™ Chemical Lactic Acid, Racemic, USP is used as a food preservative, curing agent, and flavoring agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

CAS Max % 100%
UN Number UN3265
Percent Purity 88 to 92%
Grade USP
Quantity 125 mL
Ignition Residue 0.05%
Packaging Amber Glass Bottle