missing translation for 'onlineSavingsMsg'
Learn More

Lactic acid, lithium salt, 99%, extra pure

Catalog No. AC413331000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413331000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC413331000 Supplier Thermo Scientific Chemicals Supplier No. 413331000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 867-55-0 | C3H5LiO3 | 96.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 867-55-0
Molecular Formula C3H5LiO3
Molecular Weight (g/mol) 96.01
MDL Number MFCD00065399
InChI Key GKQWYZBANWAFMQ-UHFFFAOYSA-M
Synonym lithium lactate, lithium 2-hydroxypropanoate, lactic acid, lithium salt, dl-lactic acid lithium salt, propanoic acid, 2-hydroxy-, monolithium salt, lithium 1+ ion lactate, l-+-lactic acid lithium salt, lithotab lactate, acmc-20akmj, lithium s-lactate
PubChem CID 23673452
IUPAC Name lithium;2-hydroxypropanoate
SMILES [Li+].CC(C(=O)[O-])O

Specifications

Absorbance 10% aq. soln., 10% aq. soln. at 0nm, 0.07 max. at 260nm
Color White
pH 6 to 9 (1% soln.)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass bottle
Linear Formula CH3CH(OH)CO2Li
Quantity 100 g
Beilstein 03, IV, 633
Formula Weight 96.01
Percent Purity ≥98.5%
Grade Extra Pure
Physical Form Crystalline Powder
Chemical Name or Material Lactic acid, lithium salt
Show More Show Less

RUO – Research Use Only