missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactic Acid, ACS, 85-90%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 500 mL |
| Packaging | Poly Bottle |
| Residue after Ignition | 0.0002 |
| Percent Purity | 85 to 90% |
| Grade | ACS |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.