missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, ACS, 85-90%, Spectrum™ Chemical

Catalog No. 18611176
Change view
Click to view available options
Quantity:
125 mL
500 mL
2.5 L
4 L
20 L
200 L
Packaging:
Drum
Glass Bottle
Poly Bottle
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611176 4 L Poly Bottle
18611178 125 mL Poly Bottle
18611181 500 mL Poly Bottle
18611179 2.5 L Glass Bottle
18611177 20 L Poly Drum
18611180 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18611176 Supplier Spectrum Chemical Mfg Cor Supplier No. L10054LTPL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50-21-5

Specifications

CAS Max % 1
Quantity 4 L
Packaging Poly Bottle
Residue after Ignition 0.0002
Percent Purity 85 to 90%
Grade ACS

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.