missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, 85%, FCC, 81-89%, Spectrum™ Chemical

Catalog No. 18603675
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
125 mL
20 L
200 L
4 L
4 x 4 L
500 mL
6 x 500 mL
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
Catalog No. Quantity Packaging
18603675 4 L Poly Bottle
18603672 125 mL Amber Glass Bottle
18603673 500 mL Poly Bottle
18603674 6 x 500 mL Poly Bottle
18603676 4 x 4 L Poly Bottle
18603677 20 L Poly Drum
18603678 200 L Drum
7 options

Catalog No. 18603675

Supplier: Spectrum Chemical Mfg Cor L10094LTPL

Only null left

L1009, 50-21-5, C3H6O3

Spectrum™ Chemical Lactic Acid, 85 Percent, FCC is used as a food preservative, curing agent, and flavoring agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

CAS Max % 100%
UN Number UN3265
Percent Purity 81 to 89%
Grade FCC
Quantity 4 L
Ignition Residue 0.1%
Packaging Poly Bottle