missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, 85.0-90.0%, ACS, MilliporeSigma™

Catalog No. MLX00206
Change view
Click to view available options
Quantity:
4 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MLX00206 500 mL
MLX00203 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MLX00206 Supplier Technidata Supplier No. LX00206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

Quantity 500 mL
Physical Form Liquid
Assay Percent Range 85.0 to 90.0%
Linear Formula CH3CHOHCOOH in water
Formula Weight 9.8
Grade ACS
Chemical Name or Material Lactic Acid, For ACS analysis