missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactic Acid, 10% (v/v), Ricca Chemical
Specifications
Specifications
| CAS | 50-21-5, 7732-18-5 |
| CAS Min % | 89.0, 9.0 |
| CAS Max % | 91.0, 11.0 |
| Molecular Formula | C3H6O3 |
| MDL Number | MFCD00004520 |
| Synonym | lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum |
| InChI Key | JVTAAEKCZFNVCJ-UHFFFAOYNA-N |
| SMILES | CC(O)C(O)=O |
| IUPAC Name | 2-hydroxypropanoic acid |
| Molecular Weight (g/mol) | 90.08 |
| Show More |