missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ LabX™ Density and Refractometer Express Starter Pack

Catalog No. 01915035
Change view
Click to view available options
For Use With (Equipment):
Density Meters and Refractometers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01915035 Density Meters and Refractometers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01915035 Supplier Mettler Toledo™ Supplier No. 11153130

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Connect up to three Mettler Toledo™ instruments at once to easily review results, generate reports, and export data into various formats. Includes one LiquiPhysics instrument license.

  • Enables simultaneous analyses of density, refractive index, pH and color
  • Supports full compliance with regulatory standards with automatic and secure data management, as well as full SOP user guidance
  • Avoid sample alteration between analyses due to small sample volume requirements
  • LabX organizes tasks and brings them directly to the instruments' touchscreen for the assigned lab technician to carry out
  • Completed analyses can be viewed at a glance with color-coded pass/fail information
  • Includes one LiquiPhysics instrument license

Specifications

Type LabX™ Starter Pack (Express Edition)
Display Type No Display
Description Full version of LabX for installation on a single computer
Includes 1 LiquiPhysics Instrument License
For Use With (Equipment) Density Meters and Refractometers