Learn More
Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 2L with 3 Ports, MPC Insert, MPC Body, and Luer Lock, 2D BPC
Using these BPC bags with 3 ports (MPC insert, MPC body, luer lock), simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.
Supplier: Thermo Scientific™ SH3071205
Description
Using Thermo Scientific™ Labtainer™ bags for 2 to 50L volumes, simplify cleaning and disposal and reduce cross-contamination risks and start up costs.
Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Labtainers can be customized using the industry's largest component library.
Highlights:
- 11 sizes from 50mL to 50L
- Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance and physical properties to ensure stability of contents
- Gamma-irradiation sterilized
- Convenient end-port design with a handle at the opposing end
- Space-efficient, ergonomic alternative to rigid walled carboys
- Customizable via the industry's largest library of qualified components
Recommended for:
Sampling – collection of in-process samples at a variety of bioprocessing stages
Harvest – collection and storage of bulk product from bioreactors
Storage – sterile storage of media, buffers and process intermediates
Separation – feeding and receiving of sterile liquids from separation system
Purification – buffer feeding to purification system, fraction collection and storage
Filling – sterile reservoir for filling systems
Transport & Shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities
Specifications
| Fewer resources used and less waste | |
| 29.5 cm x 31 cm (11.6 in. x 12.2 in.) | |
| 2 L, each | |
| Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.) | |
| Line 2. MPC body, polycarbonate, Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.) | |
| Edge Ports: 2 x 0.38 in. 1 x 0.25 in. |
| 2 L | |
| Packages placed in strong cardboard cartons with product and lot identification | |
| Gamma Irradiated | |
| Line 3. Body luer lock connection, polypropylene Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.) | |
| Labtainer | |
| CX5-14 |