Learn More
Thermo Scientific™ Labtainer™ BPC Bag with 2 Ports, Luer Lock and MPC Insert
Description
Using Thermo Scientific™ Labtainer™ BPC bag with 2 top ports (luer lock), simplify cleaning and disposal and reduce cross-contamination risks for 50 to 2000mL volumes of media, buffers, water for injection (WFI) and biological products.
Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Labtainers can be customized using the industry's largest component library.
Highlights:
- 11 sizes from 50mL to 50L
- Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance and physical properties to ensure stability of contents
- Gamma-irradiation sterilized
- Convenient end-port design with a handle at the opposing end
- Space-efficient, ergonomic alternative to rigid walled carboys
- Customizable via the industry's largest library of qualified components
Recommended for:
Sampling – collection of in-process samples at a variety of bioprocessing stages
Harvest – collection and storage of bulk product from bioreactors
Storage – sterile storage of media, buffers, and process intermediates
Separation – feeding and receiving of sterile liquids from separation systems
Purification – buffer feeding to purification system, fraction collection and storage
Filling – sterile reservoir for filling systems
Transport & Shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities
Specifications
Specifications
| Content And Storage | Ambient |
| Description | Two-dimensional disposable cell culture/bioprocess bag |
| Film Description | CX5-14 Single Web Film |
| Port Detail | Female Luer Lock, 12in. of 1/4in. Tubing; MPC Insert, No Tubing |
| Capacity (Metric) | 250 mL |