missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 5L with 3 Ports, MPC Insert, MPC Body, and Luer Lock, 2D BPC
GREENER_CHOICE

Catalog No. SH3071201
Change view
Click to view available options
Quantity:
5 L, each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3071201 5 L, each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3071201 Supplier Thermo Scientific™ Supplier No. SH30712.01

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Using these BPC bags with 3 ports (MPC insert, MPC body, luer lock), simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.

Using Thermo Scientific™ Labtainer™ bags for 2 to 50L volumes, simplify cleaning and disposal and reduce cross-contamination risks and start up costs.

Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Labtainers can be customized using the industry's largest component library.

Highlights:

  • 11 sizes from 50mL to 50L
  • Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance and physical properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Convenient end-port design with a handle at the opposing end
  • Space-efficient, ergonomic alternative to rigid walled carboys
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Sampling – collection of in-process samples at a variety of bioprocessing stages

  • Harvest – collection and storage of bulk product from bioreactors

  • Storage – sterile storage of media, buffers and process intermediates

  • Separation – feeding and receiving of sterile liquids from separation system

  • Purification – buffer feeding to purification system, fraction collection and storage

  • Filling – sterile reservoir for filling systems

  • Transport & Shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications

Capacity (Metric) 5 L
Dimensions 37.6 cm x 33.3 cm (14.8 in. x 13.1 in.)
Outer Packaging Packages placed in strong cardboard cartons with product and lot identification
Quantity 5 L, each
Sterilization Method Gamma Irradiated
Tubing Line 1 Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
Tubing Line 3 Line 3. Body luer lock connection, polypropylene Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Tubing Line 2 Line 2. MPC body, polycarbonate, Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.)
Product Line Labtainer
Port Detail Edge Ports: 2 x 0.38 in. 1 x 0.25 in.
Film Description CX5-14
Show More Show Less