missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 2L with 2 Ports, Luer Lock, 2D BPC
GREENER_CHOICE

Catalog No. SH3065816
Change view
Click to view available options
Tubing Style:
12 in. of 1/4 in. Tubing
No Tubing
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tubing Style
SH3065816 12 in. of 1/4 in. Tubing
SH3065716 No Tubing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3065816 Supplier Thermo Scientific™ Supplier No. SH30658.16

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Simplify cleaning and disposal and reduce cross-contamination risks for 50 to 2000mL volumes of media, buffers, water for injection (WFI) and biological products.

Using Thermo Scientific™ Labtainer™ BPC bag with 2 top ports (luer lock and MPC insert), simplify cleaning and disposal and reduce cross-contamination risks for 50 to 2000mL volumes of media, buffers, water for injection (WFI) and biological products.

Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function Labtainers can be customized using the industry's largest component library.

Highlights:

  • 11 sizes from 50mL to 50L
  • Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance, and physical properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Convenient end-port design with a handle at the opposing end
  • Space-efficient, ergonomic alternative to rigid walled carboys
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Sampling – collection of in-process samples at a variety of bioprocessing stages

  • Harvest – collection and storage of bulk product from bioreactors

  • Storage – sterile storage of media, buffers and process intermediates

  • Separation – feeding and receiving of sterile liquids from separation systems

  • Purification – buffer feeding to purification system, fraction collection and storage

  • Filling – sterile reservoir for filling systems

  • Transport &

    Shipping - bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications

Dimensions 34.8 cm x 24.4 cm (13.7 in. x 9.6 in.)
Outer Packaging Packages placed in strong cardboard cartons with product and lot identification
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity 2 L, 10 per case
Sterilization Method Gamma Irradiated
Tubing Line 1 Line 1. Body luer lock connection,polypropylene Tubing: C-Flex, length: 30 cm (12 in.) Id x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Tubing Line 2 Line 2. Insert luer lock connection, polypropylene Tubing: C-Flex, length: 30 cm (12 in.) ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Product Line Labtainer
Port Detail Edge Ports: 2 x 0.25 in.
Film Description CX5-14
Tubing Style 12 in. of 1/4 in. Tubing
Show More Show Less