missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPC insert and body connectors
GREENER_CHOICE

Catalog No. SS00105I
Change view
Click to view available options
Capacity (Metric):
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SS00105I 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SS00105I Supplier Thermo Scientific™ Supplier No. SS00105I

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 2 ports (1 MPC insert + cap, 1 MPC body + cap). It is available in 50, 100 and 200L capacities.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors

BPC port specifications:

  • Line 1: MPC insert with 9.5mm (3/8 in.) barb, cap, and pinch clamp. Tubing: Silicone, length: 51cm (20 in.), ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
  • Line 2: MPC body with 9.5mm (3/8 in.) barb, plug, and pinch clamp.

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) TK-0100-05
Includes 2 Ports (1 MPC Insert + Cap, 1 Mpc Body + Cap)
Material Polyethylene
Sterility Sterile
Tubing Style C-Flex, 51 cm (20 in.)
Film Description ASI™ 26/77
Outer Packaging 4 mil. poly bag, One 4-mil. poly bag
Port Detail MPC connections + caps, pinch clamps
Product Line Labtainer
Protective Inner Packaging Poly bags on tubing ends, protective material around rough areas
Quantity 200 L, 5 per case
Shipping Condition Room Temperature
Sterilization Method Irradiated
Dimensions (L x W x H) 142.2 x 95.3 cm (56.0 x 37.5 in.)
Length (English) 56 in.
Width (English) 37.5 in.
Product Type Bioprocess Container
Capacity (Metric) 200 L
Length (Metric) 142.2 cm
Width (Metric) 95.3 cm
Show More Show Less