missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, capsule filter, MPC insert, injection port

Description
This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 3 ports (1 membrane capsule filter, MPC insert, pinch clamps, and an injection site). It is available in 1, 5, 10 and 20L capacities.
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquid
- Collecting chromatography feed and fraction
- Harvesting, storing, and transporting bulk drug product and drug precursors
BPC port specifications:
- Line 1: 0.2μm PES membrane capsule filter + pinch clamp. Tubing: TPE 374, length: 30.5cm (12 in.), ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
- Line 2: MPC insert + cap, pinch clamp. Tubing: TPE 374, length: 91.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Includes | 3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site) |
| Material | Polyethylene |
| Tubing Style | TPE |
| Film Description | ASI™ 26/77 |
| Outer Packaging | Two 4-mil. poly bags |
| Port Detail | Capsule filter and MPC insert + cap |
| Product Line | Labtainer |
| Protective Inner Packaging | Protective material around rough areas |
| Quantity | 15/Cs. |
| Show More |