missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, capsule filter, MPC insert, injection port
GREENER_CHOICE

Catalog No. SS00159I
Change view
Click to view available options
Capacity (Metric):
1 L
5 L
10 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SS00159I 5 L
SS00158I 1 L
SS00160I 10 L
SS00161I 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SS00159I Supplier Thermo Scientific™ Supplier No. SS00159I
Only null left

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 3 ports (1 membrane capsule filter, MPC insert, pinch clamps, and an injection site). It is available in 1, 5, 10 and 20L capacities.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors

BPC port specifications:

  • Line 1: 0.2μm PES membrane capsule filter + pinch clamp. Tubing: TPE 374, length: 30.5cm (12 in.), ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
  • Line 2: MPC insert + cap, pinch clamp. Tubing: TPE 374, length: 91.

Specifications

Content And Storage Store at room temperature.
Includes 3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
Material Polyethylene
Tubing Style TPE
Film Description ASI™ 26/77
Outer Packaging Two 4-mil. poly bags
Port Detail Capsule filter and MPC insert + cap
Product Line Labtainer
Protective Inner Packaging Protective material around rough areas
Quantity 15/Cs.
Shipping Condition Room Temperature
Sterilization Method Irradiated
Dimensions (L x W x H) 32 x 31.6 cm (12.6 x 12.4 in.)
Width (English) 12.4 in.
Product Type Bioprocess Container
Capacity (Metric) 5 L
Length (Metric) 32 cm
Width (Metric) 31.6 cm
Show More Show Less