missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPC insert and body connectors, injection site
GREENER_CHOICE

Catalog No. p-7111319
Change view
Click to view available options
Capacity (Metric):
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SS00040I 10 L
SS00041I 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SS00040I Supplier Thermo Scientific™ Supplier No. SS00040I
Only null left

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 3 ports (1 MPC insert + cap, 1 MPC body + cap, 1 injection site). Available in 10L or 20L capacity.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors

BPC port specifications:

  • Line 1: MPC insert with 0.97cm (0.38 in.) barb, cap, and pinch clamp. Tubing: TPE 374, length: 30cm (12 in.), ID x OD: 9.5 x 16mm (0.38 x 0.63 in.)
  • Line 2: MPC body with 0.97cm (0.38 in.) barb, plug, and pinch c

Specifications

Content And Storage Store at room temperature.
Includes 3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
Material Polyethylene
Tubing Style TPE, 30 cm (12 in.)
Film Description ASI™ 26/77
Outer Packaging One 4-mil. poly bag
Port Detail MPC connections and injection port
Product Line Labtainer
Quantity 25/Cs.
Shipping Condition Room Temperature
Sterilization Method Irradiated
Dimensions (L x W x H) 32 x 56.5 cm (12.6 x 22.3 in.)
Width (English) 22.3 in.
Product Type Bioprocess Container
Capacity (Metric) 10 L
Length (Metric) 32 cm
Width (Metric) 56.5 cm
Show More Show Less