missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 2 ports, capsule filter and MPC insert
GREENER_CHOICE

Catalog No. SS00162I
Change view
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SS00162I 50 L
SS00163I 100 L
SS00164I 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SS00162I Supplier Thermo Scientific™ Supplier No. SS00162I

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 2 ports (1 membrane capsule filter and 1 MPC insert with pinch clamps). It is available in 50, 100 and 200L capacities.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors

BPC port specifications:

  • Line 1: 0.2μm PES membrane capsule filter + pinch clamp. Tubing: C-Flex 374, length: 91.4cm (36 in.), ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
  • Line 2: MPC insert + cap, pinch clamp. Tubing: C-Flex 374, length: 91.

Specifications

Content And Storage Store at room temperature.
Includes 2 Ports (1 Membrane Capsule Filter and 1 MPC Insert With Pinch Clamps)
Material Polyethylene
Tubing Style C-Flex, 91.4 cm (36 in.)
Film Description ASI™ 26/77
Outer Packaging Two 4-mil. poly bags
Port Detail Capsule filter and MPC insert + cap
Product Line Labtainer
Protective Inner Packaging Protective material around rough areas
Quantity 5/Cs.
Shipping Condition Room Temperature
Sterilization Method Irradiated
Dimensions (L x W x H) 95.3 x 54.6 cm (37.5 x 21.5 in.)
Length (English) 37.5 in.
Width (English) 21.5 in.
Product Type Bioprocess Container
Capacity (Metric) 50 L
Length (Metric) 95.3 cm
Width (Metric) 54.6 cm
Show More Show Less