missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 10L with 3 Ports, MPC Insert, MPC Body, and Luer Lock, 2D BPC
GREENER_CHOICE

Catalog No. SH3071202
Change view
Click to view available options
Quantity:
10 L, each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3071202 10 L, each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3071202 Supplier Thermo Scientific™ Supplier No. SH30712.02

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Using these BPC bags with 3 ports (MPC insert, MPC body, luer lock), simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.

Using Thermo Scientific™ Labtainer™ bags for 2 to 50L volumes, simplify cleaning and disposal and reduce cross-contamination risks and start up costs.

Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Labtainers can be customized using the industry's largest component library.

Highlights:

  • 11 sizes from 50mL to 50L
  • Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance and physical properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Convenient end-port design with a handle at the opposing end
  • Space-efficient, ergonomic alternative to rigid walled carboys
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Sampling – collection of in-process samples at a variety of bioprocessing stages

  • Harvest – collection and storage of bulk product from bioreactors

  • Storage – sterile storage of media, buffers and process intermediates

  • Separation – feeding and receiving of sterile liquids from separation system

  • Purification – buffer feeding to purification system, fraction collection and storage

  • Filling – sterile reservoir for filling systems

  • Transport & Shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications

Capacity (Metric) 10 L
Dimensions 62.2 cm x 30 cm (24.5 in. x 11.8 in.)
Outer Packaging Packages placed in strong cardboard cartons with product and lot identification
Quantity 10 L, each
Sterilization Method Gamma Irradiated
Tubing Line 1 Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
Tubing Line 3 Line 3. Body luer lock connection, polypropylene Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Tubing Line 2 Line 2. MPC body, polycarbonate, Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.)
Product Line Labtainer
Port Detail Edge Ports: 2 x 0.38 in. 1 x 0.25 in.
Film Description CX5-14
Show More Show Less