missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 10L with 3 Ports, MPC Insert and Injection Port, 2D BPC_
GREENER_CHOICE

Catalog No. SH3070902
Change view
Click to view available options
Quantity:
10 L, each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3070902 10 L, each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3070902 Supplier Thermo Scientific™ Supplier No. SH30709.02

Simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.

Using Thermo Scientific™ Labtainer™ BPC bag with 3 ports (MPC insert and injection), simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.

Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Labtainers can be customized using the industry's largest component library.

Highlights:

  • 11 sizes from 50mL to 50L
  • Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioproces liquids: superior stress resistance, puncture resistance and physical properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Convenient end-port design with a handle at the opposing end
  • Space-efficient, ergonomic alternative to rigid walled carboys
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Sampling – collection of in-process samples at a variety of bioprocessing stages

  • Harvest – collection and storage of bulk product from bioreactors

  • Storage – sterile storage of media, buffers and process intermediates

  • Separation – feeding and receiving of sterile liquids from separation system

  • Purification – buffer feeding to purification system, fraction collection and storage

  • Filling – sterile reservoir for filling systems

  • Transport & Shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications

Capacity (Metric) 10 L
Dimensions 62.2 cm x 30 cm (24.5 in. x 11.8 in.)
Outer Packaging Packages placed in strong cardboard cartons with product and lot identification
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity 10 L, each
Sterilization Method Gamma Irradiated
Tubing Line 1 Line 1. MPC insert, polycarbonate, No Tubing
Tubing Line 3 Line 3. Injection Port, No Tubing
Tubing Line 2 Line 2. MPC insert, polycarbonate,No Tubing
Product Line Labtainer
Port Detail Edge Ports: 2 x 0.38 in. 1 x 0.25 in.
Film Description CX5-14
Show More Show Less