missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert and body connectors, injection site
Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.
Supplier: Thermo Scientific™ SS00013I
Description
This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal originâfree ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquid
- Collecting chromatography feed and fraction
- Harvesting, storing, and transporting bulk drug product and drug precursors
Specifications
| Store at room temperature. | |
| 3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site) | |
| EVA | |
| One 4-mil. poly bag | |
| Labtainer | |
| Room Temperature | |
| 20.3 x 16.5 cm (8.0 x 6.5 in.) | |
| 20.3 cm | |
| 16.5 cm |
| 500 mL | |
| EVA | |
| ASI™ 28 | |
| MPC insert and body connectors and injection site | |
| 50/Cs. | |
| Irradiated | |
| 8 in. | |
| 6.5 in. | |
| Bioprocess Container |