missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, C-Flex 374 MPC insert and body connectors, injection site

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

Supplier:  Thermo Scientific™ SS00020I

Catalog No. SS00020I

Only null left
Explore available promotions

Description

Description

This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors
Specifications

Specifications

5 L
31.5 x 32.0 cm (12.4 x 12.6 in.)
One 4-mil. poly bag
Ethyl Vinyl Acetate
Room Temperature
Labtainer
Bioprocess Container
Store at room temperature.
ASI™ 28
3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
30/Cs.
Irradiated
C-Flex, EVA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions