missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, Luer Lock connectors, injection site
GREENER_CHOICE

Catalog No. SS00010I
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
1 L
2 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SS00010I 2 L
SS00006I 100 mL
SS00007I 250 mL
SS00009I 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SS00010I Supplier Thermo Scientific™ Supplier No. SS00010I

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors

Specifications

Content And Storage Store at room temperature.
Includes 3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
Material EVA
Tubing Style EVA
Film Description ASI™ 28
Outer Packaging One 4-mil. poly bag
Port Detail Luer lock connectors, reducer and injection site
Product Line Labtainer
Quantity 50/Cs.
Shipping Condition Room Temperature
Sterilization Method Irradiated
Dimensions (L x W x H) 33.3 x 20.3 cm (13.1 x 8.0 in.)
Width (English) 8 in.
Product Type Bioprocess Container
Capacity (Metric) 2 L
Length (Metric) 33.3 cm
Width (Metric) 20.3 cm
Show More Show Less