missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ 2D BioProcess Container, BioTitan retainers, 3 ports, Single Pack
GREENER_CHOICE

Catalog No. sh3129503
Change view
Click to view available options
Capacity (Metric):
2 L
5 L
10 L
20 L
50 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SH3129503 20 L
SH3129505 2 L
SH3129501 5 L
SH3129502 10 L
SH3129504 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. SH3129503 Supplier Thermo Scientific™ Supplier No. SH31295.03
Only null left

The Thermo Scientific 2D Labtainer™ with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.

The Thermo Scientific 2D Labtainer with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.

Features of the Thermo Scientific 2D BPC system include:

  • Now manufactured with the BioTitan Retention Device—designed to provide superior universal retention that minimizes the risk of leakage and failure at the connection point
  • Use to eliminate post-use cleaning steps required with re-useable containers and to reduce cross-contamination risks
  • Constructed in an ISO 7 cleanroom under cGMP conditions

Applications of the Thermo Scientific 2D BPC include:

  • Harvest from, feed into, and collect samples from bioreactors and fermentors
  • Buffer, culture media, and process liquid preparation and storage
  • Chromatography feed and fraction collection
  • Harvest, storage, and transport of bulk drug product and bulk drug precursors

Customize standard BPC systems for optimized single-use technology

Choose from the industry's largest component library with over 20,000 unique components that include:

  • Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
  • Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
  • Filters—used for bioburden reduction; sterilization grade filters and vent filters
  • Tubing—thermoplastic elastomers, platinum-cured silicone
  • Sample ports—septum-style and needle-free ports
  • Clamps—sliders, pinch, and crimp bands

Line 1

MPC insert, polycarbonate

Tubing length: 30 cm (12 in.)

ID x OD: 9.7 x 12.7 mm (0.38 x 0.5 in.)

Line 2

MPC insert, polycarbonate

Tubing length: 30 cm (12 in.)

ID x OD: 9.7 x 12.7 mm (0.38 x 0.5 in.)

Line 3

End plug, polypropylene

Tubing length: 8 cm (3 in.)

ID x OD: 6.35 x 10.92 mm (0.25 x 0.43 in.)

Specifications

Capacity (Metric) 20 L
Quantity 20 L
Tubing Length Line 1 30 cm (12 in.)
Tubing Length Line 3 8 cm (3 in.)
Tubing Length Line 2 30 cm (12 in.)
Tubing Material Line 3 Polypropylene
Tubing Material Line 1 Polycarbonate
Tubing Material Line 2 Polycarbonate
Tubing Style TPE