Learn More
Labplas Secure-T Sample Bags
Made to handle the most robust blending applications
Description
- Durable
- Can with stand blender use
- Contaminant-free
- Virgin polyethylene
- Large bag mouth opening
- Easy to use pull tabs
- Open top
Specifications
| 1650 mL | |
| Polyethylene | |
| 305 x 178mm (12 x 7 in.) |
| Tear-off Protection Strip | |
| 4 mil |