Learn More
Fisherbrand™ Laboratory Needle Valve
Fisherbrand™ Laboratory Needle Valves are deck mounted and intended for precision use.
Description
- Straight pattern forged brass
- Floating stainless steel needle and replaceable stainless steel seat
- Four-arm handle with color coded index disc
Specifications
| Chrome-plated Brass | |
| Needle valve assembly | |
| 15.4 cm | |
| 6.4 cm | |
| 14.0 cm |
| 1 Turret, Air Service | |
| 6.1 in. | |
| 2.5 in. | |
| 5.5 in. |