missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Tuttnauer™ Lab Line ELC-PVG-P Autoclave

Catalog No. p-7193546
Change view
Click to view available options
Capacity (Metric):
28 L
160 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
13890050 28 L
13890052 160 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13890050 Supplier Heidolph™ Supplier No. 023210683

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Autoclave offers a steam generator, vacuum pump, and cooling coils for efficient air removal and complete drying. Designed to sterilize glassware, plastics, liquids and media.

Standard Operating Features:

  • Designed to provide high quality, repeatable performance and accountability for a wide range of applications used in modern laboratories
  • For use with Agar preparation as well as the sterilization of liquids, pipettes, glasses, instruments, and biohazards with various cooling options
  • Equipped with a vacuum pump, cooling coils, and a steam generator for all laboratory sterilization needs
  • Compact size allows autoclave to fit in your lab.
  • Fast cooling (C) saves up to 75% of the cool down times of liquid samples
  • Immediate Steam (G) and Efficient Air Removal (PV) Steam generator in combination with vacuum (PV G) for complete drying
  • PID (Proportional Integral Differential) pressure control
  • Stores the last 200 cycles in built-in memory
  • Two independent, flexible PT100 temperature sensors help prevent boil over of liquids
  • Compressed air is passed through a microbiological filter into the autoclave chamber after sterilization to prevent a drop in pressure which causes load deformation, cracks, and spills
  • Cold water is circulated through cooling pipes to rapidly reduce the temperature of the chamber and liquid load to a safe temperature
  • Highly efficient drying is achieved by uniformly heating the chamber wall of the benchtop autoclave using a coiled pipe
  • The vacuum pump can be used for post vacuum drying to ensure improved drying of porous loads and hollow instruments such as pipette tips
  • Double locking safety device prevents door from opening while chamber is pressurized
  • Control lock-out switch prevents starting a cycle if door is not properly locked
  • Control system protection prevents door from opening at high pressure and temperature
  • All units include trays and printer
  • Units require utility connection to treated water, tap water, compressed air, and a floor sink

Specifications

Autoclavable Not Autoclavable
Amperage ∼16 A
Certifications/Compliance UL, cUL, ASME
Depth (Metric) Tray 41.5 cm
Depth (English) Tray 16.3 in.
Capacity (English) 7 gal.
Capacity (Metric) 28 L
Depth (Metric) Chamber 45 cm
Diameter (English) Chamber 11 in.
Depth (English) Chamber 18 in.
Diameter (Metric) Chamber 28 cm
For Use With (Application) Sterilization
For Use With (Equipment) Liquids, Glassware, Utensils, Wrapped, Unwrapped, and Solids
Length (English) 29.9 in.
Height (English) Exterior 17.3 in.
Height (Metric) Exterior 44 cm
Length (Metric) 76 cm
Model 2840 EL-PV-GC
Product Type Autoclave
Temperature (English) 221°F to 278°F
Temperature (Metric) 105°C to 137°C
Display Multi-color to display the stage of the cycle
Net Weight (English) 94.8 lb.
Net Weight (Metric) 43 kg
Printer Included
Voltage 230 V
Wattage 2200 w
Width (English) Tray 6.7 in.
Width (Metric) Tray 17 cm
Sterility Non-sterile
Shipping Weight (Metric) 76.2 kg
Dimensions (L x W x H) 20.9 x 24.8 x 17.3 in. (53 x 63 x 44 cm)
Show More Show Less