Learn More
Labomed™ 2.5X Infinity Plan Objective
Ideal for pathology applications.
Supplier: Labo America Inc 9142002
Description
- Must be used with a flip-top condenser supplied standard with Non-Phase Lx 500 configurations
Specifications
| Infinity Plan Objective | |
| Pathology applications | |
| Lx 500 |