missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ LabKlenz 200 Acid Detergent

Catalog No. 17467592
Change view
Click to view available options
Size:
1 gal.
5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17467592 5 gal.
17467591 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17467592 Supplier STERIS™ Supplier No. 1L2005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

LabKlenz 200 Acid Detergent: Low-foaming, phosphoric acid-based liquid for effective removal of lab residues. Environmentally friendly and reliable. pH (1%): 2.0.

LabKlenz 200 Detergent is specially formulated for use in automated glassware washing applications both as a rinse aid and for the removal of particulate, scale, and inorganic soils.

HOW LABKLENZ 200 DETERGENT WORKS

LabKlenz 200 Detergent is a low-foaming, phosphoric acid based liquid detergent.

WHY LABKLENZ 200 DETERGENT?

  • Flexibility - Effective removal of laboratory residues in both soft and hard water
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - Formulated with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility

Physical Properties

pH (1%): 2.0, typical

Specifications

Product Type Detergent
Color-coded No
Form Clear Liquid
pH 2.0
Scent Unscented
Sterility Non-sterile
Container Type Pail
Size 5 gal.
Volume (Metric) 18.93 L