missing translation for 'onlineSavingsMsg'
Learn More

STERIS LabKlenz™ 100 Alkaline Detergent

Alkaline detergent with corrosion inhibitors

Supplier:  STERIS 1L1005

 View more versions of this product

Catalog No. 29-636-6303


Only null left
Explore available promotions

Description

Description

  • Formulated to meet unique cleaning and materials compatibility requirements found in laboratory glassware washing applications
  • High alkaline for effective removal of residues in both soft and hard water
  • Contains corrosion inhibitors for excellent materials compatibility
  • Reduces utility consumption and cleaning time with low foaming, free-rinsing formulation
  • Phosphate-free formulation with biodegradable surfactants
  • Manufactured in cGMP compliant, FDA registered and ISO 9001 certified facility
  • Specific gravity: 1.15
Specifications

Specifications

Non-sterile
Certificate of analysis
Laboratory washers
Yellow
Plastic jerrican
5 gal.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions