missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Minigrip™ LabGuard™ Biohazard Specimen Bags
Provide safe and secure transport of multiple samples
$208.13
Specifications
| Closure Type | Ziplock |
|---|---|
| Color | Clear |
| For Use With (Application) | Provide safe and secure transport of multiple samples |
| Includes | Absorbent pad, reclosable zipper, biohazard symbol and document pouch |
| Material | Plastic |
| Catalog Number | Mfr. No. | Height (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Height (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
|
22311203
|
Minigrip™
SBL2AP69B |
22.86 cm | 15.2 cm |
Case of 1000 Each for $208.13
|
|
||||
Description
Provide safe and secure transport of multiple samples
- Three walls
- With reclosable zipper
- Thickness of 1.75 mil
Specifications
| Ziplock | |
| Provide safe and secure transport of multiple samples | |
| Plastic | |
| 1.75 mil |
| Clear | |
| Absorbent pad, reclosable zipper, biohazard symbol and document pouch | |
| 50 Pack | |
| 3-wall Biohazard Specimen Bag |